missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxycarbonylethyl)benzeneboronic acid, 97%

Catalog No. p-7034843
Change view
Click to view available options
Quantity:
250 mg
1 g
Catalog No. Quantity
AAH52542MD 250 mg
AAH5254203 1 g
2 options

Catalog No. AAH52542MD

Supplier: Thermo Scientific Chemicals H52542MD

Only null left

CAS: 833472-82-5 | C10H13BO4 | 208.02 g/mol

3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 833472-82-5
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD04115648
InChI Key IPXZIWCEVNDHFD-UHFFFAOYSA-N
Synonym 3-2-methoxycarbonylethyl phenylboronic acid, methyl 3-3-boronophenyl propionate, 3-3-methoxy-3-oxopropyl phenyl boronic acid, 3-3-methoxy-3-oxopropyl phenylboronic acid, 3-2-methoxycarbonylethyl phenyl boronic acid, acmc-209prx, benzenepropanoic acid,3-borono-, 1-methyl ester
PubChem CID 4993880
IUPAC Name [3-(3-methoxy-3-oxopropyl)phenyl]boronic acid
SMILES COC(=O)CCC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 96°C to 100°C
Quantity 250 mg
Formula Weight 208.02
Percent Purity 97%
Chemical Name or Material 3-(2-Methoxycarbonylethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only