missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxy-3-pyridyl)acrylic acid, 95%

Catalog No. AAH5006303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5006303 1 g
AAH5006306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5006303 Supplier Thermo Scientific Chemicals Supplier No. H5006303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 131674-41-4 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131674-41-4
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD07781156
InChI Key MTVRDAQJMCORAQ-SNAWJCMRSA-N
Synonym 3-2-methoxy-3-pyridyl acrylic acid, e-3-2-methoxypyridin-3-yl acrylic acid, 3-2-methoxypyridin-3-yl acrylic acid, 2e-3-2-methoxypyridin-3-yl prop-2-enoic acid, 2e-3-2-methoxy 3-pyridyl prop-2-enoic acid
PubChem CID 14691926
IUPAC Name (E)-3-(2-methoxypyridin-3-yl)prop-2-enoic acid
SMILES COC1=C(C=CC=N1)C=CC(=O)O

Specifications

Quantity 1 g
Formula Weight 179.18
Percent Purity 95%
Chemical Name or Material 3-(2-Methoxy-3-pyridyl)acrylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only