missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC46109DA

Quantity 1g

 View more versions of this product

Catalog No. CC46109DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

89929-93-1
174.199
SBTRFADZILHXNG-UHFFFAOYSA-N
11116545
C1=CC(=CC(=C1)CO)C2=CC=CO2
C11H10O2
MFCD06203100
3-2-furyl phenyl methanol, 3-furan-2-yl phenyl methanol, 3-2-furyl benzyl alcohol, 3-2-furyl benzenemethanol, 3-2-furanyl phenylmethanol, 3-2-furanyl-phenylmethanol, 3-2-furanyl phenyl methanol, 3-2-furyl phenyl methan-1-ol
[3-(furan-2-yl)phenyl]methanol

Specifications

[3-(2-Furyl)phenyl]methanol
94.5
95%
C11H10O2
MFCD06203100
SBTRFADZILHXNG-UHFFFAOYSA-N
[3-(furan-2-yl)phenyl]methanol
11116545
95%
89929-93-1
100.0
Amber Glass Bottle
1g
3-2-furyl phenyl methanol, 3-furan-2-yl phenyl methanol, 3-2-furyl benzyl alcohol, 3-2-furyl benzenemethanol, 3-2-furanyl phenylmethanol, 3-2-furanyl-phenylmethanol, 3-2-furanyl phenyl methanol, 3-2-furyl phenyl methan-1-ol
C1=CC(=CC(=C1)CO)C2=CC=CO2
174.199
174.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents