missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC46114DE

Quantity 5g

 View more versions of this product

Catalog No. CC46114DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

102269-42-1
159.188
SLNKACMTMZYMNA-UHFFFAOYSA-N
4161309
C1=CC(=CC(=C1)N)C2=CC=CO2
C10H9NO
MFCD04039071
3-furan-2-yl aniline, 3-2-furyl aniline, benzenamine,3-2-furanyl, acmc-1c42n, 3-2-furyl aniline hydrochloride
3-(furan-2-yl)aniline

Specifications

3-(2-Furyl)aniline
96.5
97%
5g
3-furan-2-yl aniline, 3-2-furyl aniline, benzenamine,3-2-furanyl, acmc-1c42n, 3-2-furyl aniline hydrochloride
C1=CC(=CC(=C1)N)C2=CC=CO2
159.188
159.19
102269-42-1
100.0
C10H9NO
MFCD04039071
SLNKACMTMZYMNA-UHFFFAOYSA-N
3-(furan-2-yl)aniline
4161309
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Promotions

Promotions