missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)acrylic Acid 98.0+%, TCI America™

Catalog No. p-7129490
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F008425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F008425G Supplier TCI America Supplier No. F008425G

Chemical Identifiers

CAS 539-47-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003257,MFCD11656544
InChI Key ZCJLOOJRNPHKAV-ONEGZZNKSA-N
Synonym 3-2-furyl acrylic acid, e-3-furan-2-yl acrylic acid, 2-furanacrylic acid, 3-furan-2-yl acrylic acid, furanacrylic acid, 2-furalacetic acid, furacrylic acid, furylacrylic acid, 2e-3-furan-2-yl prop-2-enoic acid, 2-propenoic acid, 3-2-furanyl
PubChem CID 643402
IUPAC Name (2E)-3-(furan-2-yl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CO1

Specifications

Melting Point 141°C
Color Yellow
Boiling Point 286°C
Quantity 25 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Furyl)acrylic Acid
EINECSNumber (9)-1158
RTECSNumber LT8560000
TSCA Yes