missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC47009DA

Quantity 1g

 View more versions of this product

Catalog No. CC47009DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

886851-33-8
178.191
3-2-furyl-1-methyl-1h-pyrazol-5-yl methanol, 5-furan-2-yl-2-methylpyrazol-3-yl methanol, 3-2-furyl-1-methylpyrazol-5-yl methan-1-ol, 3-furan-2-yl-1-methyl-1h-pyrazol-5-yl methanol
[5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
C9H10N2O2
BYXYWFXMSJTOKB-UHFFFAOYSA-N
18525804
CN1C(=CC(=N1)C2=CC=CO2)CO

Specifications

[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol
97%
1g
BYXYWFXMSJTOKB-UHFFFAOYSA-N
[5-(furan-2-yl)-2-methylpyrazol-3-yl]methanol
18525804
97%
886851-33-8
C9H10N2O2
3-2-furyl-1-methyl-1h-pyrazol-5-yl methanol, 5-furan-2-yl-2-methylpyrazol-3-yl methanol, 3-2-furyl-1-methylpyrazol-5-yl methan-1-ol, 3-furan-2-yl-1-methyl-1h-pyrazol-5-yl methanol
CN1C(=CC(=N1)C2=CC=CO2)CO
178.191
178.19
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents