missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Fluorophenyl)propionic acid, 96%

Catalog No. p-7032593
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3411003 1 g
AAH3411009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3411003 Supplier Thermo Scientific Chemicals Supplier No. H3411003
Only null left

CAS: 1643-26-1 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1643-26-1
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD01310820
InChI Key GUZLQEOSDXLCKX-UHFFFAOYSA-N
PubChem CID 2063866
IUPAC Name 3-(2-fluorophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CCC(=O)O)F

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
Synonym 3-2-fluorophenyl propionic acid, 3-2-fluorophenyl propanoic acid, benzenepropanoic acid, 2-fluoro, 3-2-fluorophenyl proprionic acid, 3-2-fluoro-phenyl-propionic acid, 2-fluorohydrocinnamic acid, acmc-1bz7v, 2'-fluorocinnamide, 2-fluoro-benzenepropanoic acid, benzenepropanoic acid,2-fluoro
Formula Weight 168.17
Percent Purity 96%
Chemical Name or Material 3-(2-Fluorophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only