missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. AAH6635006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6635006 5 g
AAH6635003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6635006 Supplier Thermo Scientific Chemicals Supplier No. H6635006

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 935854-36-7
MDL Number MFCD22493823
Synonym Ethyl 3-(3-[4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl]phenyl)propionate

Specifications

Color White to Yellow
Molecular Formula C17H25BO4
Quantity 5 g
Beilstein 20464729
Solubility Information Insoluble in water
Formula Weight 304.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-[2-(Ethoxycarbonyl)ethyl]benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only