missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid, 98%

CAS: 23598-72-3 | C11H8ClNO3 | 237.64 g/mol

Supplier:  Thermo Scientific Chemicals L0624030

 View more versions of this product

Catalog No. AAL0624030


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
23598-72-3
C11H8ClNO3
791928
UVEPOHNXGXVOJE-UHFFFAOYSA-N
3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
90203
≥98%
187°C to 192°C
250 g
MFCD00020813
3-2-chlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2'-chlorophenyl-4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl, 3-2-chloro-phenyl-5-methyl-isoxazole-4-carboxylic acid, 3-2-chlorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-o-chlorophenyl-5-methyl, 4-carboxy-3-2-chlorophenyl-5-methylisoxazole, 3 2-chlorophenyl-5-methyl-isoxazolyl-4-carboxylic acid, 5-methyl-3-2-chlorophenyl-4-isoxazolecarboxylic acid
CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl
237.64
237.64

Chemical Identifiers

23598-72-3
237.64
UVEPOHNXGXVOJE-UHFFFAOYSA-N
90203
CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl
C11H8ClNO3
MFCD00020813
3-2-chlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2'-chlorophenyl-4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl, 3-2-chloro-phenyl-5-methyl-isoxazole-4-carboxylic acid, 3-2-chlorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-o-chlorophenyl-5-methyl, 4-carboxy-3-2-chlorophenyl-5-methylisoxazole, 3 2-chlorophenyl-5-methyl-isoxazolyl-4-carboxylic acid, 5-methyl-3-2-chlorophenyl-4-isoxazolecarboxylic acid
3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 245-770-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only