Learn More
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid, 98%
CAS: 23598-72-3 | C11H8ClNO3 | 237.64 g/mol
Supplier: Thermo Scientific Chemicals L0624018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid | |
| 23598-72-3 | |
| C11H8ClNO3 | |
| 791928 | |
| UVEPOHNXGXVOJE-UHFFFAOYSA-N | |
| 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid | |
| 90203 | |
| ≥98% |
| 187°C to 192°C | |
| 50 g | |
| MFCD00020813 | |
| 3-2-chlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2'-chlorophenyl-4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl, 3-2-chloro-phenyl-5-methyl-isoxazole-4-carboxylic acid, 3-2-chlorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-o-chlorophenyl-5-methyl, 4-carboxy-3-2-chlorophenyl-5-methylisoxazole, 3 2-chlorophenyl-5-methyl-isoxazolyl-4-carboxylic acid, 5-methyl-3-2-chlorophenyl-4-isoxazolecarboxylic acid | |
| CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl | |
| 237.64 | |
| 237.64 |
Chemical Identifiers
| 23598-72-3 | |
| 237.64 | |
| UVEPOHNXGXVOJE-UHFFFAOYSA-N | |
| 90203 | |
| CC1=C(C(O)=O)C(=NO1)C1=CC=CC=C1Cl |
| C11H8ClNO3 | |
| MFCD00020813 | |
| 3-2-chlorophenyl-5-methylisoxazole-4-carboxylic acid, 5-methyl-3-2'-chlorophenyl-4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3-2-chlorophenyl-5-methyl, 3-2-chloro-phenyl-5-methyl-isoxazole-4-carboxylic acid, 3-2-chlorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-o-chlorophenyl-5-methyl, 4-carboxy-3-2-chlorophenyl-5-methylisoxazole, 3 2-chlorophenyl-5-methyl-isoxazolyl-4-carboxylic acid, 5-methyl-3-2-chlorophenyl-4-isoxazolecarboxylic acid | |
| 3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 245-770-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only