missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one 98.0+%, TCI America™

Catalog No. p-7125458
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24335G 5 g
C243325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24335G Supplier TCI America Supplier No. C24335G
Only null left

Chemical Identifiers

CAS 63234-80-0
Molecular Formula C11H15ClN2O
Molecular Weight (g/mol) 226.704
MDL Number MFCD06200813
InChI Key CMWCQQUYLPYOMY-UHFFFAOYSA-N
PubChem CID 2763079
IUPAC Name 3-(2-chloroethyl)-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2CCCCC2=N1)CCCl

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.70
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one