missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Bromophenyl)propionic Acid, TCI America™

$27.50 - $91.67

Chemical Identifiers

CAS 15115-58-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD01310791
InChI Key AOACQJFIGWNQBC-UHFFFAOYSA-N
Synonym 3-2-bromophenyl propionic acid, 3-2-bromophenyl propanoic acid, benzenepropanoic acid, 2-bromo, 3-2-bromophenyl propionicacid, 3-2-bromo-phenyl-propionic acid, r-2-bromo-3-benzenepropanoic acid, pubchem15787, acmc-20a0iu, intermediates-zcf02152, 2-bromobenzenepropionic acid
PubChem CID 316010
IUPAC Name 3-(2-bromophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CCC(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44095G View Documents TCI America
B44095G
5g Each for $27.50
 
B440925G View Documents TCI America
B440925G
25g Each for $91.67
 
Specifications

Chemical Identifiers

15115-58-9
229.073
AOACQJFIGWNQBC-UHFFFAOYSA-N
316010
C1=CC=C(C(=C1)CCC(=O)O)Br
C9H9BrO2
MFCD01310791
3-2-bromophenyl propionic acid, 3-2-bromophenyl propanoic acid, benzenepropanoic acid, 2-bromo, 3-2-bromophenyl propionicacid, 3-2-bromo-phenyl-propionic acid, r-2-bromo-3-benzenepropanoic acid, pubchem15787, acmc-20a0iu, intermediates-zcf02152, 2-bromobenzenepropionic acid
3-(2-bromophenyl)propanoic acid

Specifications

186°C
99°C
15115-58-9
MFCD01310791
AOACQJFIGWNQBC-UHFFFAOYSA-N
3-(2-bromophenyl)propanoic acid
316010
Crystal-Powder at 20°C
3-(2-Bromophenyl)propionic Acid
Yellow
5g
C9H9BrO2
3-2-bromophenyl propionic acid, 3-2-bromophenyl propanoic acid, benzenepropanoic acid, 2-bromo, 3-2-bromophenyl propionicacid, 3-2-bromo-phenyl-propionic acid, r-2-bromo-3-benzenepropanoic acid, pubchem15787, acmc-20a0iu, intermediates-zcf02152, 2-bromobenzenepropionic acid
C1=CC=C(C(=C1)CCC(=O)O)Br
229.073
229.07
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No