missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Boc-amino)ethyl]azetidine, 95%

Catalog No. AAH63797MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63797MD 250 mg
AAH6379703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63797MD Supplier Thermo Scientific Chemicals Supplier No. H63797MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 162696-31-3 | C10H20N2O2 | 200.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162696-31-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD09878605
InChI Key CSPJUZSNJFANNU-UHFFFAOYSA-N
Synonym 3-boc-aminoethylazetidine, 3-n-boc-aminoethyl azetidine, tert-butyl 2-azetidin-3-yl ethylcarbamate, tert-butyl 2-azetidin-3-yl ethyl carbamate, tert-butyl n-2-azetidin-3-yl ethyl carbamate, 3-2-n-boc-aminoethyl azetidine, tert-butyl 2-3-azetidinyl ethylcarbamate, 2-azetin-3-yl-1-n-boc ethylamine, tert-butyl 2-azetidin-3-yl ethylcarbamate hydrochloride, carbamic acid, n-2-3-azetidinyl ethyl-, 1,1-dimethylethyl ester
PubChem CID 37818431
IUPAC Name tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCC1CNC1

Specifications

Melting Point 84°C to 86°C
Color Brown to White
Quantity 250 mg
Formula Weight 200.28
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-[2-(Boc-amino)ethyl]azetidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only