missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. AAH5170906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5170906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5170906 Supplier Thermo Scientific Chemicals Supplier No. H5170906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1257651-17-4 | Molecular Formula: C19H29BN2O5 | Molecular Weight: 376.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1257651-17-4
Molecular Formula C19H29BN2O5
Molecular Weight (g/mol) 376.26
MDL Number MFCD16294500
InChI Key IYTPRTWBNLJZLK-UHFFFAOYSA-N
Synonym 3-2-boc-amino acetamido benzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl-carbamic acid tert-butyl ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamoyl methyl carbamate, amtb154, 3-2-boc-amino acetamido phenylboronic acid pinacol ester, 3-2-tert-butoxycarbonylamino acetamido phenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl carbamic acid tert-butyl ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylcarbamoyl methyl carbamic acid tert-butyl ester, tert-butyl 2-oxo-2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anilino ethyl carbamate
PubChem CID 57415693
IUPAC Name tert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 5 g
Formula Weight 376.26
Percent Purity 97%
Chemical Name or Material 3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only