missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Benzothiazolyl)-7-(diethylamino)coumarin 98.0+%, TCI America™

Catalog No. p-7119472
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B20881G 1 g
B20885G 5 g
2 options

Catalog No. B20881G

Supplier: TCI America B20881G

Chemical Identifiers

CAS 38215-36-0
Molecular Formula C20H18N2O2S
Molecular Weight (g/mol) 350.436
MDL Number MFCD00041869
InChI Key VBVAVBCYMYWNOU-UHFFFAOYSA-N
Synonym coumarin 6, 3-2-benzothiazolyl-7-diethylamino coumarin, 2h-1-benzopyran-2-one, 3-2-benzothiazolyl-7-diethylamino, 3-benzo d thiazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-1,3-benzothiazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-1,3-benzothiazol-2-yl-7-diethylamino chromen-2-one, 3-2-benzthiazolyl-7-diethylaminocoumarin, 3-2-benzothiazolyl-n,n-diethylumbelliferylamine, 3-benzothiazol-2-yl-7-diethylamino chromen-2-one, coumarin-6
PubChem CID 100334
ChEBI CHEBI:51942
IUPAC Name 3-(1,3-benzothiazol-2-yl)-7-(diethylamino)chromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4S3

Specifications

Melting Point 208°C
Quantity 1 g
Formula Weight 350.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Benzothiazolyl)-7-(diethylamino)coumarin