missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin 98.0+%, TCI America™
$63.38 - $372.81
Chemical Identifiers
| CAS | 27425-55-4 |
|---|---|
| Molecular Formula | C20H19N3O2 |
| Molecular Weight (g/mol) | 333.391 |
| MDL Number | MFCD00051348 |
| InChI Key | GOLORTLGFDVFDW-UHFFFAOYSA-N |
| Synonym | coumarin 7, 3-2-benzimidazolyl-7-diethylamino coumarin, 2h-1-benzopyran-2-one, 3-1h-benzimidazol-2-yl-7-diethylamino, disperse yellow 82, coumarin, 3-2-benzimidazolyl-7-diethylamino, 3-1h-benzimidazol-2-yl-7-diethylamino-2-benzopyrone, 3-1h-benzimidazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-1h-benzo d imidazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-benzimidazol-2-yl-7-diethylaminocoumarin, 3-1h-benzimidazol-2-yl-7-diethylamino chromen-2-one |
| PubChem CID | 94381 |
| IUPAC Name | 3-(1H-benzimidazol-2-yl)-7-(diethylamino)chromen-2-one |
| SMILES | CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4N3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2111100MG
|
TCI America
B2111100MG |
100 mg |
Each for $63.38
|
|
|||||
|
B21111G
|
TCI America
B21111G |
1 g |
Each for $372.81
|
|
|||||
Chemical Identifiers
| 27425-55-4 | |
| 333.391 | |
| GOLORTLGFDVFDW-UHFFFAOYSA-N | |
| 94381 | |
| CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4N3 |
| C20H19N3O2 | |
| MFCD00051348 | |
| coumarin 7, 3-2-benzimidazolyl-7-diethylamino coumarin, 2h-1-benzopyran-2-one, 3-1h-benzimidazol-2-yl-7-diethylamino, disperse yellow 82, coumarin, 3-2-benzimidazolyl-7-diethylamino, 3-1h-benzimidazol-2-yl-7-diethylamino-2-benzopyrone, 3-1h-benzimidazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-1h-benzo d imidazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-benzimidazol-2-yl-7-diethylaminocoumarin, 3-1h-benzimidazol-2-yl-7-diethylamino chromen-2-one | |
| 3-(1H-benzimidazol-2-yl)-7-(diethylamino)chromen-2-one |
Specifications
| 27425-55-4 | |
| C20H19N3O2 | |
| 100 mg | |
| GOLORTLGFDVFDW-UHFFFAOYSA-N | |
| 3-(1H-benzimidazol-2-yl)-7-(diethylamino)chromen-2-one | |
| 94381 | |
| ≥98.0% (HPLC,T) | |
| 3-(2-Benzimidazolyl)-7-(diethylamino)coumarin |
| 242°C | |
| MFCD00051348 | |
| coumarin 7, 3-2-benzimidazolyl-7-diethylamino coumarin, 2h-1-benzopyran-2-one, 3-1h-benzimidazol-2-yl-7-diethylamino, disperse yellow 82, coumarin, 3-2-benzimidazolyl-7-diethylamino, 3-1h-benzimidazol-2-yl-7-diethylamino-2-benzopyrone, 3-1h-benzimidazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-1h-benzo d imidazol-2-yl-7-diethylamino-2h-chromen-2-one, 3-benzimidazol-2-yl-7-diethylaminocoumarin, 3-1h-benzimidazol-2-yl-7-diethylamino chromen-2-one | |
| CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4N3 | |
| 333.391 | |
| 333.39 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0677
TSCA : Yes