missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 242°C |
| Quantity | 100 mg |
| Formula Weight | 333.39 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(2-Benzimidazolyl)-7-(diethylamino)coumarin |
Safety and Handling
| EINECSNumber | (5)-0677 |
| TSCA | Yes |