missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%

Catalog No. AAB2315914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2315914 25 g
AAB2315922 100 g
AAB2315936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2315914 Supplier Thermo Scientific Chemicals Supplier No. B2315914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3069-29-2 | C8H22N2O2Si | 206.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3069-29-2
Molecular Formula C8H22N2O2Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00054776
InChI Key MQWFLKHKWJMCEN-UHFFFAOYSA-N
Synonym n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
PubChem CID 76482
IUPAC Name N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine
SMILES CO[Si](C)(CCCNCCN)OC

Specifications

Boiling Point 265°C
Quantity 25 g
UN Number UN2735
Beilstein 2074259
Flash Point 90°C (194°F)
Formula Weight 206.36
Percent Purity 97%
Odor Amine-like
Refractive Index 1.449
Sensitivity Air and moisture sensitive
Density 0.968
Chemical Name or Material 3-(2-Aminoethylamino)propylmethyldimethoxysilane
Show More Show Less
Hazard Category H227-H302-H315-H317-H318
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 221-336-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only