missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Aminoethylamino)propyldimethoxymethylsilane 97.0+%, TCI America™
$69.31 - $531.34
Chemical Identifiers
| CAS | 3069-29-2 |
|---|---|
| Molecular Formula | C8H22N2O2Si |
| Molecular Weight (g/mol) | 206.36 |
| MDL Number | MFCD00054776 |
| InChI Key | MQWFLKHKWJMCEN-UHFFFAOYSA-N |
| Synonym | n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl |
| PubChem CID | 76482 |
| IUPAC Name | (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine |
| SMILES | CO[Si](C)(CCCNCCN)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A087625ML
|
TCI America
A087625ML |
25 mL |
Each for $69.31
|
|
|||||
|
A0876500ML
|
TCI America
A0876500ML |
500 mL |
Each for $531.34
|
|
|||||
Chemical Identifiers
| 3069-29-2 | |
| 206.36 | |
| MQWFLKHKWJMCEN-UHFFFAOYSA-N | |
| 76482 | |
| CO[Si](C)(CCCNCCN)OC |
| C8H22N2O2Si | |
| MFCD00054776 | |
| n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl | |
| (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine |
Specifications
| 3069-29-2 | |
| 139°C | |
| MFCD00054776 | |
| 3267 | |
| MQWFLKHKWJMCEN-UHFFFAOYSA-N | |
| (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine | |
| 76482 | |
| ≥97.0% (GC,T) | |
| 3-(2-Aminoethylamino)propyldimethoxymethylsilane |
| Yellow | |
| C8H22N2O2Si | |
| 25 mL | |
| n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl | |
| CO[Si](C)(CCCNCCN)OC | |
| 206.36 | |
| 206.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2084
TSCA : Yes