missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyldimethoxymethylsilane 97.0+%, TCI America™

Catalog No. A087625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A087625ML 25 mL
A0876500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A087625ML Supplier TCI America Supplier No. A087625ML

Chemical Identifiers

CAS 3069-29-2
Molecular Formula C8H22N2O2Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00054776
InChI Key MQWFLKHKWJMCEN-UHFFFAOYSA-N
Synonym n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
PubChem CID 76482
IUPAC Name (2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine
SMILES CO[Si](C)(CCCNCCN)OC

Specifications

Color Yellow
Boiling Point 139°C
Quantity 25 mL
UN Number 3267
Formula Weight 206.36
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(2-Aminoethylamino)propyldimethoxymethylsilane
EINECSNumber (2)-2084
TSCA Yes