missing translation for 'onlineSavingsMsg'
Learn More

3-(2,5-Dimethoxyphenyl)propionic acid, 96%

Catalog No. aah3356606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3356606 5 g
AAH3356603 1 g
AAH3356614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3356606 Supplier Thermo Scientific Chemicals Supplier No. H3356606
Only null left

CAS: 10538-49-5 | C11H14O4 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10538-49-5
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00060325
InChI Key JENQUCZZZGYHRW-UHFFFAOYSA-N
PubChem CID 66343
IUPAC Name 3-(2,5-dimethoxyphenyl)propanoic acid
SMILES COC1=CC=C(OC)C(CCC(O)=O)=C1

Specifications

Melting Point 61°C to 66°C
Quantity 5 g
Synonym 3-2,5-dimethoxyphenyl propanoic acid, 3-2,5-dimethoxyphenyl propionic acid, benzenepropanoic acid, 2,5-dimethoxy, 2,5-dimethoxyhydrocinnamic acid, 2,5-dimethoxybenzenepropanoic acid, benzenepropanoic acid,2,5-dimethoxy, maybridge1_000808, acmc-1bo6n, ksc503q0f, 2,5-dimethoxy-benzenepropanoic acid
Formula Weight 210.23
Percent Purity 96%
Chemical Name or Material 3-(2,5-Dimethoxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only