missing translation for 'onlineSavingsMsg'
Learn More

3-(2,4-Difluorobenzoyl)propionic Acid 98.0+%, TCI America™

Catalog No. D45851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45851G 1 g
D45855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45851G Supplier TCI America Supplier No. D45851G

Chemical Identifiers

CAS 110931-77-6
Molecular Formula C10H8F2O3
Molecular Weight (g/mol) 214.17
MDL Number MFCD00143016
InChI Key OKYUHFSCTFNDFB-UHFFFAOYSA-N
Synonym 4-(2,4-Difluorophenyl)-4-oxobutyric Acid
PubChem CID 2774081
IUPAC Name 4-(2,4-difluorophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)C1=C(F)C=C(F)C=C1

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Formula Weight 214.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2,4-Difluorobenzoyl)propionic Acid