missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

Catalog No. p-2698869
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC21901CB 250mg
CC21901DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC21901CB Supplier Thermo Scientific Chemicals Supplier No. CC21901CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61471-45-2
Molecular Formula C11H8NO2
Molecular Weight (g/mol) 186.19
MDL Number MFCD02656610
InChI Key PODFNQCZFHLJPH-UHFFFAOYSA-M
Synonym 3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
PubChem CID 736537
IUPAC Name 3-(1H-pyrrol-1-yl)benzoate
SMILES [O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)benzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.