missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC21901DA

Quantity 1g

 View more versions of this product

Catalog No. CC21901DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

61471-45-2
186.19
PODFNQCZFHLJPH-UHFFFAOYSA-M
736537
[O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1
C11H8NO2
MFCD02656610
3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
3-pyrrol-1-ylbenzoic acid

Specifications

3-(1H-Pyrrol-1-yl)benzoic acid
96.5
97%
C11H8NO2
MFCD02656610
PODFNQCZFHLJPH-UHFFFAOYSA-M
3-pyrrol-1-ylbenzoic acid
736537
97%
61471-45-2
100.0
Amber glass bottle
1g
3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
[O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1
186.19
187.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents