missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-3-yl)benzoic acid, 97%, Thermo Scientific™

Catalog No. p-4510472
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31701DA 1g Amber Glass Bottle
CC31701DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31701DA Supplier Thermo Scientific Chemicals Supplier No. CC31701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 850375-11-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD03407956
InChI Key RXZRBZWATRFHCS-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-5-yl benzoic acid, 3-1h-pyrazol-3-yl benzoic acid, 3-2h-pyrazol-3-yl benzoic acid, benzoic acid, 3-1h-pyrazol-3-yl, 5-3-carboxyphenyl-1h-pyrazole, pubchem22748, buttpark 44\01-87, d0tx9p, 3-pyrazol-3-ylbenzoic acid, 3-pyrazol-5-ylbenzoic acid
PubChem CID 2795542
IUPAC Name 3-(1H-pyrazol-5-yl)benzoic acid
SMILES OC(=O)C1=CC=CC(=C1)C1=CC=NN1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-3-yl)benzoic acid