missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-yl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC32904DE
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC32904DE 5g
CC32904CB 250mg
CC32904DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC32904DE Supplier Thermo Scientific Chemicals Supplier No. CC32904DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 852227-92-0
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD06740321
InChI Key NKFXXJOWQSOGOF-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-yl benzaldehyde, 3-1-pyrazolyl benzaldehyde, 3-1h-pyrazol-1-yl benzenecarbaldehyde, 3-pyrazol-1-yl-benzaldehyde, 3-pyrazol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrazol-1-yl, 3-pyrazolylbenzaldehyde, benzaldehyde, 3-1h-pyrazol-1-yl
PubChem CID 7172309
IUPAC Name 3-pyrazol-1-ylbenzaldehyde
SMILES C1=CC(=CC(=C1)N2C=CC=N2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5g
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 3-(1H-pyrazol-1-yl)benzaldehyde
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.