missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-yl)benzaldehyde, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC32904DA

Quantity 1g

 View more versions of this product

Catalog No. CC32904DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

852227-92-0
172.187
NKFXXJOWQSOGOF-UHFFFAOYSA-N
7172309
C1=CC(=CC(=C1)N2C=CC=N2)C=O
C10H8N2O
MFCD06740321
3-1h-pyrazol-1-yl benzaldehyde, 3-1-pyrazolyl benzaldehyde, 3-1h-pyrazol-1-yl benzenecarbaldehyde, 3-pyrazol-1-yl-benzaldehyde, 3-pyrazol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrazol-1-yl, 3-pyrazolylbenzaldehyde, benzaldehyde, 3-1h-pyrazol-1-yl
3-pyrazol-1-ylbenzaldehyde

Specifications

3-(1H-pyrazol-1-yl)benzaldehyde
96.5
97%
C10H8N2O
MFCD06740321
NKFXXJOWQSOGOF-UHFFFAOYSA-N
3-pyrazol-1-ylbenzaldehyde
7172309
97%
852227-92-0
100.0
Amber glass bottle
1g
3-1h-pyrazol-1-yl benzaldehyde, 3-1-pyrazolyl benzaldehyde, 3-1h-pyrazol-1-yl benzenecarbaldehyde, 3-pyrazol-1-yl-benzaldehyde, 3-pyrazol-1-yl benzaldehyde, benzaldehyde,3-1h-pyrazol-1-yl, 3-pyrazolylbenzaldehyde, benzaldehyde, 3-1h-pyrazol-1-yl
C1=CC(=CC(=C1)N2C=CC=N2)C=O
172.187
172.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents