missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC43204CB
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC43204CB 250mg
CC43204DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC43204CB Supplier Thermo Scientific Chemicals Supplier No. CC43204CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102432-05-3
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD08271917
InChI Key JMKQKKQFLYPQDD-UHFFFAOYSA-N
Synonym 3-1h-imidazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-imidazol-1-ylmethyl, 3-imidazol-1-ylmethyl benzaldehyde, 3-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde, 3-1h-imidazol-1-ylmethyl, acmc-1c5ro, 3-imidazolylmethyl benzaldehyde
PubChem CID 10397537
IUPAC Name 3-(imidazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)CN2C=CN=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 3-(1H-imidazol-1-ylmethyl)benzaldehyde
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.