missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Imidazol-1-ylmethyl)aniline, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Quantity | 250mg |
| Formula Weight | 173.22 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-(1H-Imidazol-1-ylmethyl)aniline |