missing translation for 'onlineSavingsMsg'
Learn More

3-(1h-1,2,4-Triazol-1-yl)aniline, 90%, Thermo Scientific™

$208.15 - $208.15

Chemical Identifiers

CAS 176032-78-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
InChI Key QOHJDIAWBVPZFL-UHFFFAOYSA-N
PubChem CID 23503656
IUPAC Name 3-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC(=C1)N)N2C=NC=N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC38414CB
SDS View Documents
Thermo Scientific Chemicals
CC38414CB
250 mg Amber Glass Bottle
Each for $208.15
Only null left
 
Description

Description

Specifications

Chemical Identifiers

176032-78-3
160.18
23503656
C1=CC(=CC(=C1)N)N2C=NC=N2
C8H8N4
QOHJDIAWBVPZFL-UHFFFAOYSA-N
3-(1,2,4-triazol-1-yl)aniline

Specifications

176032-78-3
Amber Glass Bottle
250 mg
QOHJDIAWBVPZFL-UHFFFAOYSA-N
3-(1,2,4-triazol-1-yl)aniline
23503656
90%
90%
C8H8N4
3-1h-1,2,4-triazol-1-yl aniline, 3-1,2,4-triazol-1-yl aniline, benzenamine,3-1h-1,2,4-triazol-1-yl, benzenamine, 3-1h-1,2,4-triazol-1-yl, 1-3-aminophenyl-1h-1,2,4-triazole, 3-1,2,4-triazolyl phenylamine, 3-1,2,4 triazol-1-yl-phenylamine, 3-1,2,4 triazol-1-yl phenylamine
C1=CC(=CC(=C1)N)N2C=NC=N2
160.18
160.18
3-(1H-1, 2, 4-Triazol-1-yl)aniline
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'