missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperazinylmethyl)indole, 95%

Catalog No. AAH63402MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63402MD 250 mg
AAH6340203 1 g
AAH6340206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63402MD Supplier Thermo Scientific Chemicals Supplier No. H63402MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 114746-66-6 | C13H17N3 | 215.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114746-66-6
Molecular Formula C13H17N3
Molecular Weight (g/mol) 215.3
InChI Key ZTNAIHGOFCMOPJ-UHFFFAOYSA-N
Synonym 3-piperazin-1-ylmethyl-1h-indole, 1h-indole,3-1-piperazinylmethyl, acmc-1c6nr, 1-1h-indole-3-ylmethyl piperazine, 1h-indole, 3-1-piperazinylmethyl-, hydrochloride
PubChem CID 17880880
IUPAC Name 3-(piperazin-1-ylmethyl)-1H-indole
SMILES C1CN(CCN1)CC2=CNC3=CC=CC=C32

Specifications

Quantity 250 mg
Formula Weight 215.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1-Piperazinylmethyl)indole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only