missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)propionic acid, 96%

Catalog No. p-7032369
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3379506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3379506 Supplier Thermo Scientific Chemicals Supplier No. H3379506

CAS: 3243-42-3 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3243-42-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00033058
InChI Key PRLKVVMRQFFIOQ-UHFFFAOYSA-N
Synonym 3-1-naphthyl propionic acid, 3-naphthalen-1-yl propanoic acid, 1-naphthalenepropanoic acid, 1-naphthalenepropionic acid, 3-1-naphthyl-propionic acid, 3-1-naphthyl propanoic acid, 3-naphth-1-yl propanoic acid, 3-naphthalen-1-yl-propionic acid, 1-naphthalenepropanoicacid, 3-naphthylpropanoic acid
PubChem CID 95254
IUPAC Name 3-naphthalen-1-ylpropanoic acid
SMILES OC(=O)CCC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 155°C to 159°C
Quantity 5 g
Formula Weight 200.24
Percent Purity 96%
Chemical Name or Material 3-(1-Naphthyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only