missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile, 97%, Thermo Scientific™

Catalog No. CC60516DE
Change view
Click to view available options
Amber Glass Bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC60516DE Amber Glass Bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC60516DE Supplier Thermo Scientific Chemicals Supplier No. CC60516DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 179055-95-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H9N3
Quantity 5g
MDL Number MFCD09702401
Synonym 3-1-methyl-1h-pyrazol-5-yl benzonitrile, 3-2-methylpyrazol-3-yl benzonitrile, benzonitrile,3-1-methyl-1h-pyrazol-5-yl, benzonitrile, 3-1-methyl-1h-pyrazol-5-yl, 3-1-methylpyrazol-5-yl benzenecarbonitrile
InChI Key NEFOVTAGRMOIIV-UHFFFAOYSA-N
SMILES CN1N=CC=C1C1=CC=CC(=C1)C#N
IUPAC Name 3-(2-methylpyrazol-3-yl)benzonitrile
Molecular Weight (g/mol) 183.21
PubChem CID 10511671
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-5-yl)benzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.