missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC60501DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC60501DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

628297-55-2
202.213
OBOMYSVFLSMYLV-UHFFFAOYSA-N
18548847
CN1C(=CC=N1)C2=CC(=CC=C2)C(=O)O
C11H10N2O2
MFCD09702398
3-1-methyl-1h-pyrazol-5-yl benzoic acid, 3-2-methylpyrazol-3-yl benzoic acid, 3-1-methylpyrazol-5-yl benzoic acid, benzoicacid,3-1-methyl-1h-pyrazol-5-yl, benzoicacid, 3-1-methyl-1h-pyrazol-5-yl, benzoic acid, 3-1-methyl-1h-pyrazol-5-yl
3-(2-methylpyrazol-3-yl)benzoic acid

Specifications

3-(1-Methyl-1H-pyrazol-5-yl)benzoic acid
96.5
97%
C11H10N2O2
MFCD09702398
OBOMYSVFLSMYLV-UHFFFAOYSA-N
3-(2-methylpyrazol-3-yl)benzoic acid
18548847
97%
628297-55-2
100.0
Amber Glass Bottle
1g
3-1-methyl-1h-pyrazol-5-yl benzoic acid, 3-2-methylpyrazol-3-yl benzoic acid, 3-1-methylpyrazol-5-yl benzoic acid, benzoicacid,3-1-methyl-1h-pyrazol-5-yl, benzoicacid, 3-1-methyl-1h-pyrazol-5-yl, benzoic acid, 3-1-methyl-1h-pyrazol-5-yl
CN1C(=CC=N1)C2=CC(=CC=C2)C(=O)O
202.213
202.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents