Learn More
3-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™
$197.13 - $197.13
Chemical Identifiers
| CAS | 912569-59-6 |
|---|---|
| Molecular Formula | C10H9ClN2O2S |
| Molecular Weight (g/mol) | 256.704 |
| MDL Number | MFCD09817503 |
| InChI Key | QGUULJKRHYTFTP-UHFFFAOYSA-N |
| PubChem CID | 24229628 |
| IUPAC Name | 3-(1-methylpyrazol-3-yl)benzenesulfonyl chloride |
| SMILES | CN1C=CC(=N1)C2=CC(=CC=C2)S(=O)(=O)Cl |
Chemical Identifiers
| 912569-59-6 | |
| 256.704 | |
| QGUULJKRHYTFTP-UHFFFAOYSA-N | |
| 3-(1-methylpyrazol-3-yl)benzenesulfonyl chloride |
| C10H9ClN2O2S | |
| MFCD09817503 | |
| 24229628 | |
| CN1C=CC(=N1)C2=CC(=CC=C2)S(=O)(=O)Cl |
Specifications
| 912569-59-6 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD09817503 | |
| 3-1-methyl-1h-pyrazol-3-yl benzenesulfonyl chloride, benzenesulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl benzenesulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl benzenesulphonyl chloride, 3-1-methyl-1h-pyrazol-3-yl benzene-1-sulfonyl chloride, chloro 3-1-methylpyrazol-3-yl phenyl sulfone, 3-1-methyl-1h-pyrazol-3-yl phenylsulfonyl chloride, 3-1-methyl-1h-pyrazol-3-yl-benzenesulfonyl chloride, benzenesulfonyl chloride,3-1-methyl-1h-pyrazol-3-yl | |
| CN1C=CC(=N1)C2=CC(=CC=C2)S(=O)(=O)Cl | |
| 256.704 | |
| 256.71 | |
| 3-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride |
| 96.5 | |
| 97% | |
| C10H9ClN2O2S | |
| 250 mg | |
| QGUULJKRHYTFTP-UHFFFAOYSA-N | |
| 3-(1-methylpyrazol-3-yl)benzenesulfonyl chloride | |
| 24229628 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger