missing translation for 'onlineSavingsMsg'
Learn More

[3-(1,3-Thiazol-2-yl)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC36509DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC36509DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

184847-97-0
191.248
VDFDKGHCZKGOLZ-UHFFFAOYSA-N
18525726
C1=CC(=CC(=C1)CO)C2=NC=CS2
C10H9NOS
MFCD09025828
3-1,3-thiazol-2-yl phenyl methanol, 3-thiazol-2-yl benzyl alcohol, 3-thiazol-2-yl phenyl methanol, 3-1,3-thiazol-2-yl phenyl methan-1-ol
[3-(1,3-thiazol-2-yl)phenyl]methanol

Specifications

[3-(1, 3-Thiazol-2-yl)phenyl]methanol
96.5
97%
C10H9NOS
MFCD09025828
VDFDKGHCZKGOLZ-UHFFFAOYSA-N
[3-(1,3-thiazol-2-yl)phenyl]methanol
18525726
97%
184847-97-0
100.0
Amber Glass Bottle
1g
3-1,3-thiazol-2-yl phenyl methanol, 3-thiazol-2-yl benzyl alcohol, 3-thiazol-2-yl phenyl methanol, 3-1,3-thiazol-2-yl phenyl methan-1-ol
C1=CC(=CC(=C1)CO)C2=NC=CS2
191.248
191.25
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents