missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dithian-2-ylidene)-2,4-pentanedione 98.0+%, TCI America™

Catalog No. D42081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42081G 1 g
D42085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42081G Supplier TCI America Supplier No. D42081G

Chemical Identifiers

CAS 55727-23-6
Molecular Formula C9H12O2S2
Molecular Weight (g/mol) 216.313
MDL Number MFCD01096981
InChI Key PGPIRMKKHQOWIK-UHFFFAOYSA-N
PubChem CID 681994
IUPAC Name 3-(1,3-dithian-2-ylidene)pentane-2,4-dione
SMILES CC(=O)C(=C1SCCCS1)C(=O)C

Specifications

Melting Point 103°C
Color Yellow
Quantity 1 g
Formula Weight 216.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(1,3-Dithian-2-ylidene)-2,4-pentanedione