missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine 95.0+%, TCI America™
$51.26 - $149.98
Chemical Identifiers
| CAS | 104372-31-8 |
|---|---|
| Molecular Formula | C10H15NO3S |
| Molecular Weight (g/mol) | 229.294 |
| MDL Number | MFCD00075428 |
| InChI Key | GBBJBUGPGFNISJ-HPFPYREMSA-N |
| Synonym | 1r---10-camphorsulfonyl oxaziridine, --2s,8ar-camphorylsulfonyl oxaziridine |
| PubChem CID | 98044902 |
| SMILES | CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C |
Chemical Identifiers
| 104372-31-8 | |
| 229.294 | |
| GBBJBUGPGFNISJ-HPFPYREMSA-N | |
| 98044902 |
| C10H15NO3S | |
| MFCD00075428 | |
| 1r---10-camphorsulfonyl oxaziridine, --2s,8ar-camphorylsulfonyl oxaziridine | |
| CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C |
Specifications
| 104372-31-8 | |
| White | |
| MFCD00075428 | |
| 1r---10-camphorsulfonyl oxaziridine, --2s,8ar-camphorylsulfonyl oxaziridine | |
| CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C | |
| 98044902 | |
| ≥95.0% (T) | |
| (2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine [Asymmetric Oxidizing Reagent] |
| 168°C | |
| C10H15NO3S | |
| 1 g | |
| GBBJBUGPGFNISJ-HPFPYREMSA-N | |
| 229.294 | |
| 229.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No