missing translation for 'onlineSavingsMsg'
Learn More

(2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine 95.0+%, TCI America™

Catalog No. C13275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13275G 5 g
C13271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13275G Supplier TCI America Supplier No. C13275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104372-31-8
Molecular Formula C10H15NO3S
Molecular Weight (g/mol) 229.294
MDL Number MFCD00075428
InChI Key GBBJBUGPGFNISJ-HPFPYREMSA-N
Synonym 1r---10-camphorsulfonyl oxaziridine, --2s,8ar-camphorylsulfonyl oxaziridine
PubChem CID 98044902
SMILES CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 229.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine [Asymmetric Oxidizing Reagent]