missing translation for 'onlineSavingsMsg'
Learn More

(2S,5S)-2,5-Hexanediol 98.0+%, TCI America™

Catalog No. p-7130832
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09691G 1 g
H09695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09691G Supplier TCI America Supplier No. H09691G

Chemical Identifiers

CAS 34338-96-0
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00082583
InChI Key OHMBHFSEKCCCBW-WDSKDSINSA-N
Synonym 2s,5s-hexane-2,5-diol, 2s,5s-2,5-hexanediol, 2s,5s-+-hexanediol, 2s,5s-2,5-dihydroxyhexane, 2s,5s-hexanediol, 2s,5s-+-2,5-hexanediol, 2,5-hexanediol, 2s,5s, pubchem6345, 2s,5s hexanediol, +-2,5-hexanediol
PubChem CID 6950288
IUPAC Name (2S,5S)-hexane-2,5-diol
SMILES C[C@H](O)CC[C@H](C)O

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 71°C
Quantity 1 g
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,5S)-2,5-Hexanediol
EINECSNumber (2)-0240
TSCA No