missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 695816-47-8 |
|---|---|
| Molecular Formula | C15H21O2P |
| Molecular Weight (g/mol) | 264.3 |
| InChI Key | JAKXTLMNIHPLEU-RYUDHWBXSA-N |
| Synonym | 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min |
| PubChem CID | 16218307 |
| IUPAC Name | 2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane |
| SMILES | CC1CCC(P1C2=CC=CC=C2C3OCCO3)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC435171000
|
Thermo Scientific Chemicals
435171000 |
100mg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 695816-47-8 | |
| 264.3 | |
| 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min | |
| 2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane |
| C15H21O2P | |
| JAKXTLMNIHPLEU-RYUDHWBXSA-N | |
| 16218307 | |
| CC1CCC(P1C2=CC=CC=C2C3OCCO3)C |
Specifications
| 695816-47-8 | |
| 100.0 | |
| >110°C | |
| 97% | |
| C15H21O2P | |
| 1.103 | |
| JAKXTLMNIHPLEU-RYUDHWBXSA-N | |
| 2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane | |
| 16218307 | |
| 97% |
| 96.0 | |
| 1.1030g/mL | |
| Authentic | |
| Glass bottle | |
| 100mg | |
| 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min | |
| CC1CCC(P1C2=CC=CC=C2C3OCCO3)C | |
| 264.3 | |
| 264.3 | |
| (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane |
RUO â Research Use Only