missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 97%, Thermo Scientific™
Specifications
Specifications
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Quantity | 100mg |
| Formula Weight | 292.35 |
| Percent Purity | 97% |
| Chemical Name or Material | (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane |
RUO – Research Use Only