missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid 97.0+%, TCI America™
Supplier: TCI America B31781G
Specifications
| (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid | |
| 162°C | |
| C10H16FNO4 | |
| 1 g | |
| YGWZXQOYEBWUTH-BQBZGAKWSA-N | |
| (2S,4S)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 2756017 | |
| ≥97.0% (GC,T) |
| 203866-13-1 | |
| White-Yellow | |
| MFCD04973957 | |
| n-boc-cis-4-fluoro-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, 2s,4s-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, n-tert-butoxycarbonyl-cis-4-fluoro-l-proline, boc-cis-4-fluoro-pro-oh, 2s,4s-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-flp-oh, boc-cis-pro 4-f-oh, boc-cis-4-fluoro-l-proline, 2s,4s-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F | |
| 233.239 | |
| 233.24 | |
| Crystalline Powder |
Chemical Identifiers
| 203866-13-1 | |
| 233.239 | |
| YGWZXQOYEBWUTH-BQBZGAKWSA-N | |
| 2756017 | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F |
| C10H16FNO4 | |
| MFCD04973957 | |
| n-boc-cis-4-fluoro-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, 2s,4s-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, n-tert-butoxycarbonyl-cis-4-fluoro-l-proline, boc-cis-4-fluoro-pro-oh, 2s,4s-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-flp-oh, boc-cis-pro 4-f-oh, boc-cis-4-fluoro-l-proline, 2s,4s-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid | |
| (2S,4S)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Safety and Handling
TSCA : No