missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid 97.0+%, TCI America™

Catalog No. B3178200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3178200MG 200 mg
B31781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3178200MG Supplier TCI America Supplier No. B3178200MG

Chemical Identifiers

CAS 203866-13-1
Molecular Formula C10H16FNO4
Molecular Weight (g/mol) 233.239
MDL Number MFCD04973957
InChI Key YGWZXQOYEBWUTH-BQBZGAKWSA-N
Synonym n-boc-cis-4-fluoro-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-fluoropyrrolidine-2-carboxylic acid, 2s,4s-1-tert-butoxycarbonyl-4-fluoro-2-pyrrolidinecarboxylic acid, n-tert-butoxycarbonyl-cis-4-fluoro-l-proline, boc-cis-4-fluoro-pro-oh, 2s,4s-1-tert-butoxy carbonyl-4-fluoropyrrolidine-2-carboxylic acid, boc-flp-oh, boc-cis-pro 4-f-oh, boc-cis-4-fluoro-l-proline, 2s,4s-1-boc-4-fluoro-2-pyrrolidinecarboxylic acid
PubChem CID 2756017
IUPAC Name (2S,4S)-4-fluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)F

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 200 mg
Formula Weight 233.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic Acid