missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(-)-Bis(diphenylphosphino)butane 93.0+%, TCI America™

Catalog No. B27041G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27041G 1 g
B2704100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27041G Supplier TCI America Supplier No. B27041G
Only null left

Chemical Identifiers

CAS 64896-28-2
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00009836
InChI Key FWXAUDSWDBGCMN-ZBXWWWQKNA-N
Synonym s,s-chiraphos, 2s,3s---bis diphenylphosphino butane, chiraphos, chiraphos, s,s, unii-6qr78gzl9b, 6qr78gzl9b, 2s,3s-butane-2,3-diylbis diphenylphosphine, phosphine, 1s,2s-1,2-dimethyl-1,2-ethanediyl bis diphenyl, --chiraphos, 2s,3s-chiraphos
PubChem CID 10113249
IUPAC Name [(2S,3S)-3-(diphenylphosphanyl)butan-2-yl]diphenylphosphane
SMILES C[C@@H]([C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 109°C
Color White
Quantity 1 g
Formula Weight 426.48
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2S,3S)-(-)-Bis(diphenylphosphino)butane