missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S,5R,6R)-5,6-Bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane 97.0+%, TCI America™

Catalog No. b3694100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3694100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3694100MG Supplier TCI America Supplier No. B3694100MG
Only null left

Chemical Identifiers

CAS 1585236-34-5
Molecular Formula C10H18N6O4
Molecular Weight (g/mol) 286.292
InChI Key VKZQUTPIKFBBMZ-IMSYWVGJSA-N
PubChem CID 53384441
IUPAC Name (2S,3S,5R,6R)-5,6-bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane
SMILES CC1(C(OC(C(O1)CN=[N+]=[N-])CN=[N+]=[N-])(C)OC)OC

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 100 mg
Physical Form Crystal-Powder at 20°C
UN Number 1325
Formula Weight 286.29
Percent Purity ≥97.0% (GC)
Chemical Name or Material (2S,3S,5R,6R)-5,6-Bis(azidomethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane