missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,3S)-3-(tert-Butoxycarbonylamino)-1,2-epoxy-4-phenylbutane 98.0+%, TCI America™
$93.21 - $186.41
Chemical Identifiers
| CAS | 98737-29-2 |
|---|---|
| Molecular Formula | C15H21NO3 |
| Molecular Weight (g/mol) | 263.337 |
| MDL Number | MFCD02258997 |
| InChI Key | NVPOUMXZERMIJK-QWHCGFSZSA-N |
| Synonym | 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b |
| PubChem CID | 9903372 |
| IUPAC Name | tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 |
Chemical Identifiers
| 98737-29-2 | |
| 263.337 | |
| NVPOUMXZERMIJK-QWHCGFSZSA-N | |
| 9903372 | |
| CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 |
| C15H21NO3 | |
| MFCD02258997 | |
| 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b | |
| tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate |
Specifications
| 98737-29-2 | |
| White-Yellow | |
| MFCD02258997 | |
| 3077 | |
| NVPOUMXZERMIJK-QWHCGFSZSA-N | |
| tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate | |
| 9903372 | |
| ≥98.0% (GC) | |
| (2S,3S)-3-(tert-Butoxycarbonylamino)-1,2-epoxy-4-phenylbutane |
| 127°C | |
| C15H21NO3 | |
| 1 g | |
| 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b | |
| CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2 | |
| 263.337 | |
| 263.34 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6533
TSCA : No