missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%

Catalog No. AC395020250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395020250 25 g Glass bottle
AC395020010 1 g Glass bottle
AC395020050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC395020250 Supplier Thermo Scientific Chemicals Supplier No. 395020250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98737-29-2 | C15H21NO3 | 263.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98737-29-2
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.34
MDL Number MFCD02258997
InChI Key NVPOUMXZERMIJK-QWHCGFSZSA-N
Synonym 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b
PubChem CID 9903372
IUPAC Name tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 125.0°C to 127.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: <0.1% (20°C)
Formula Weight 263.34
Percent Purity 98%
Chemical Name or Material (2S, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only