missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,3S)-3-Methylpyrrolidine-2-carboxylic acid, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Beige |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Specific Rotation Condition | − 28.00 (25.00°C c=0.7,water) |
| Specific Rotation | − 28.00 |
| Formula Weight | 129.16 |
| Percent Purity | 97% |
| Physical Form | Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
RUO – Research Use Only