Learn More
(2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%
CAS: 73397-27-0 | C12H23NO5 | 261.32 g/mol
Supplier: Thermo Scientific Chemicals H5256903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (2S,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid | |
| C12H23NO5 | |
| 1 g | |
| DJZCWTDKDFJARG-UHFFFAOYNA-N | |
| (2S,3S)-2-hydroxy-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 2762291 | |
| 97% |
| 73397-27-0 | |
| MFCD04974450 | |
| 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, n-boc-2s,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2s,3s-2-hydroxy-5-methyl-3-2-methylpropan-2-yl oxycarbonylamino hexanoic acid, 2s,3s-3-t-butoxycarbonyl amino-2-hydroxy-5-methylhexaneoic acid, 2s,3s-2-hydroxy-3-tert-butyloxycarbonyl amino-5-methylhexanoic acid, 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoicacid, hexanoic acid,3-1,1-dimethylethoxy carbonyl amino-2-hydroxy-5-methyl-, 2s,3s | |
| CC(C)CC(NC(=O)OC(C)(C)C)C(O)C(O)=O | |
| 261.32 | |
| 261.32 |
Chemical Identifiers
| 73397-27-0 | |
| 261.32 | |
| DJZCWTDKDFJARG-UHFFFAOYNA-N | |
| 2762291 |
| C12H23NO5 | |
| MFCD04974450 | |
| 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, n-boc-2s,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2s,3s-2-hydroxy-5-methyl-3-2-methylpropan-2-yl oxycarbonylamino hexanoic acid, 2s,3s-3-t-butoxycarbonyl amino-2-hydroxy-5-methylhexaneoic acid, 2s,3s-2-hydroxy-3-tert-butyloxycarbonyl amino-5-methylhexanoic acid, 2s,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoicacid, hexanoic acid,3-1,1-dimethylethoxy carbonyl amino-2-hydroxy-5-methyl-, 2s,3s | |
| CC(C)CC(NC(=O)OC(C)(C)C)C(O)C(O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only